9,9-Diphenyl-9,10-dihydroacridine ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
9,10-Dihydro-9,9-diphenylacridine
D5332-200MG
D5332-1G
TCIAD5332-200MGEA
80
EUR
TCIAD5332-200MG
TCIAD5332-1G
9,9-Diphenyl-9,10-dihydroacridine ≥98.0% (by HPLC, total nitrogen)
9,9-Diphenyl-9,10-dihydroacridine
Formula:
C₂₅H₁₉N MW: 333,43 g/mol |
Número MDL:
MFCD28127357 Núm. CAS: 20474-15-1 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 249.0 to 254.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...