Dimethyl 3,5-Pyridinedicarboxylate ≥98.0% (by GC, titration analysis)
Proveedor: TCI
D5364-25G
D5364-5G
TCIAD5364-25GEA
214
EUR
TCIAD5364-25G
TCIAD5364-5G
Dimethyl 3,5-Pyridinedicarboxylate ≥98.0% (by GC, titration analysis)
Dimethyl 3,5-Pyridinedicarboxylate
Formula:
C₉H₉NO₄ Pto. Ebullición: 132°C |
Número MDL:
MFCD00518827 Núm. CAS: 4591-55-3 EINECS: 224-981-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...