Dimethyl iminodiacetate hydrochloride 98,0 Titrimetric

Proveedor: TCI
Atención

Sinónimos: Dimethyl 2,2'-azanediyldiacetate hydrochloride

D5385-5G D5385-25G
TCIAD5385-5GEA 55 EUR
TCIAD5385-5G TCIAD5385-25G
Dimethyl iminodiacetate hydrochloride 98,0 Titrimetric
Dimethyl iminodiacetate hydrochloride
Formula: C₆H₁₁NO₄·HCl
MW: 197.62 g/mol
Número MDL: MFCD00216724
Núm. CAS: 39987-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40d95707d1f07aeb00ea74dd7d51e7057e5299d5&vid=40d95707d1f07aeb00ea74dd7d51e7057e5299d5&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR