9,9'-Diethyl-9H,9'H-3,3'-bicarbazole ≥97.0% (by total nitrogen basis)

Proveedor: TCI
Atención

D5387-1G D5387-5G
TCIAD5387-1GEA 82 EUR
TCIAD5387-1G TCIAD5387-5G
9,9'-Diethyl-9H,9'H-3,3'-bicarbazole ≥97.0% (by total nitrogen basis)
9,9'-Diethyl-9H,9'H-3,3'-bicarbazole
Formula: C₂₈H₂₄N₂
MW: 388,51 g/mol
Número MDL: MFCD32062883
Núm. CAS: 20466-00-6

Order Now

Resultados test especificaciones

Appearance White to Amber powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 190.0 to 194.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR