3,6-Dibromo-9-(3,5-dimethylphenyl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

D5421-200MG D5421-1G
TCIAD5421-200MGEA 86 EUR
TCIAD5421-200MG TCIAD5421-1G
3,6-Dibromo-9-(3,5-dimethylphenyl)-9H-carbazole ≥97.0% (by HPLC, total nitrogen)
3,6-Dibromo-9-(3,5-dimethylphenyl)-9H-carbazole
Formula: C₂₀H₁₅Br₂N
MW: 429,16 g/mol
Número MDL: MFCD31618134
Núm. CAS: 1873364-08-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ac0aaa57c10200f0dbbf361cacd86a0ed439862&vid=6ac0aaa57c10200f0dbbf361cacd86a0ed439862&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 239.0 to 243.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR