2-Chloropyridine-3,4-diamine ≥96.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
3,4-Diamino-2-chloropyridine, 2-Chloropyridine-3,4-diamine
D5455-1G
D5455-5G
TCIAD5455-1GEA
69
EUR
TCIAD5455-1G
TCIAD5455-5G
2-Chloropyridine-3,4-diamine ≥96.0% (by GC, titration analysis)
2-Chloropyridine-3,4-diamine
Formula:
C₅H₆ClN₃ MW: 143,5742 g/mol |
Número MDL:
MFCD07363739 Núm. CAS: 39217-08-8 UN: 2811 ADR: 6.1,III |
Resultados test especificaciones
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.000 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...