3,3'-Di-tert-pentyl-4H,4'H-[1,1'-binaphthalenylidene]-4,4'-dione ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
2-(1,1-Dimethylpropyl)-4-[3-(1,1-dimethylpropyl)-4-oxo-1(4H)-naphthalenylidene]-1(4H)-naphthalenone, 3,3'-Di-tert-amyl-4H,4'H-[1,1'-binaphthalenylidene]-4,4'-dione
D5506-1G
D5506-5G
TCIAD5506-1GEA
31
EUR
TCIAD5506-1G
TCIAD5506-5G
3,3'-Di-tert-pentyl-4H,4'H-[1,1'-binaphthalenylidene]-4,4'-dione ≥98.0% (by HPLC)
3,3'-Di-tert-pentyl-4H,4'H-[1,1'-binaphthalenylidene]-4,4'-dione
Formula:
C₃₀H₃₂O₂ MW: 424,58 g/mol |
Número MDL:
MFCD28045103 Núm. CAS: 334634-19-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 197.0 to 201.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...