Dodecylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

Sinónimos: Dodecylammonium bromide, Laurylamine hydrobromide, Dodecan-1-amine hydrobromide, 1-Aminododecane hydrobromide

D5537-1G D5537-5G
TCIAD5537-1GEA 25 EUR
TCIAD5537-1G TCIAD5537-5G
Dodecylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Dodecylamine hydrobromide
Formula: C₁₂H₂₇N·HBr
MW: 266,27 g/mol
Pto. de fusión: 190°C
Número MDL: MFCD30543762
Núm. CAS: 26204-55-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
water max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR