Dichloroglyoxime 98,0 N, T

Proveedor: TCI

D5562-1G D5562-5G
TCIAD5562-1GEA 119 EUR
TCIAD5562-1G TCIAD5562-5G
Dichloroglyoxime 98,0 N, T
Dichloroglyoxime
Formula: C₂H₂Cl₂N₂O₂
MW: 156.95 g/mol
Número MDL: MFCD01326506
Núm. CAS: 2038-44-0

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point (Decomposition) 200.0 to 204.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR