N-[4-(2,4-Dimethylphenyl)-2-thiazolyl]-4-pyridinecarboxamide ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: N-[4-(2,4-Dimethylphenyl)thiazol-2-yl]isonicotinamide, Nek2/Hec1 inhibitor

D5563-10MG D5563-50MG
TCIAD5563-10MGEA 98 EUR
TCIAD5563-10MG TCIAD5563-50MG
N-[4-(2,4-Dimethylphenyl)-2-thiazolyl]-4-pyridinecarboxamide ≥98.0% (by HPLC)
N-[4-(2,4-Dimethylphenyl)-2-thiazolyl]-4-pyridinecarboxamide
Formula: C₁₇H₁₅N₃OS
MW: 309,39 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD03384899
Núm. CAS: 560103-80-2

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 13.00 to 14.20 %
Melting point 147.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR