N,9-Diphenyl-9H-carbazol-3-amine ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

Sinónimos: 3-(N-Phenylamino)-9-phenyl-9H-carbazole

D5567-1G D5567-5G
TCIAD5567-1GEA 64 EUR
TCIAD5567-1G TCIAD5567-5G
N,9-Diphenyl-9H-carbazol-3-amine ≥98.0% (by HPLC, total nitrogen)
N,9-Diphenyl-9H-carbazol-3-amine
Formula: C₂₄H₁₈N₂
MW: 334,42 g/mol
Número MDL: MFCD26401776
Núm. CAS: 894791-43-6

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR