N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-diphenyl-9H-fluoren-2-amine ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
D5588-1G
D5588-5G
TCIAD5588-1GEA
105
EUR
TCIAD5588-1G
TCIAD5588-5G
N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-diphenyl-9H-fluoren-2-amine ≥98.0% (by HPLC, total nitrogen)
N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-diphenyl-9H-fluoren-2-amine
Formula:
C₄₀H₃₁N MW: 525,70 g/mol |
Número MDL:
MFCD32062889 Núm. CAS: 1456702-56-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 244.0 to 248.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...