N,N-Dimethyl-1,3-propanediamine dihydroiodide ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
N,N-Dimethyl-1,3-propanediammonium Diiodide, N1,N1-Dimethylpropane-1,3-diamine dihydroiodide, 3-(Dimethylamino)propylamine dihydroiodide
D5619-1G
D5619-5G
TCIAD5619-1GEA
29
EUR
TCIAD5619-1G
TCIAD5619-5G
N,N-Dimethyl-1,3-propanediamine dihydroiodide ≥97.0% (by titrimetric analysis)
N,N-Dimethyl-1,3-propanediamine dihydroiodide
Formula:
C₅H₁₄N₂·₂HI |
Núm. CAS:
2561497-43-4 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...