Diphemanil Methylsulphate ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-(Diphenylmethylene)-1,1-dimethylpiperidin-1-ium methyl sulphate, 4-(Diphenylmethylene)-1,1-dimethylpiperidin-1-ium Methyl sulphate, Diphenmanil methylsulphate
D5623-1G
D5623-250MG
TCIAD5623-1GEA
697
EUR
TCIAD5623-1G
TCIAD5623-250MG
Diphemanil Methylsulphate ≥98.0% (by HPLC, total nitrogen)
Diphemanil Methylsulphate
Formula:
C₂₁H₂₇NO₄OS MW: 405,51 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00865157 Núm. CAS: 62-97-5 EINECS: 200-552-4 Índice Merck: 14,03306 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 192.0 to 196.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...