trans-4-Dimethylaminocrotonic acid hydrochloride ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(E)-4-Dimethylaminocrotonic acid hydrochloride, (E)-4-(Dimethylamino)-2-butenoic acid hydrochloride, (E)-4-(Dimethylamino)but-2-enoic acid hydrochloride
D5625-1G
D5625-5G
TCIAD5625-1GEA
34
EUR
TCIAD5625-1G
TCIAD5625-5G
trans-4-Dimethylaminocrotonic acid hydrochloride ≥98.0% (by HPLC, titration analysis)
trans-4-Dimethylaminocrotonic acid hydrochloride
Formula:
C₆H₁₂ClNO₂ MW: 165,62 g/mol |
Número MDL:
MFCD03695466 Núm. CAS: 848133-35-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 160.0 to 164.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...