3,4-Dimethyl-1H-pyrazole Phosphate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3,4-Dimethyl-1H-pyrazole phosphate
D5668-25G
D5668-5G
TCIAD5668-25GEA
132
EUR
TCIAD5668-25G
TCIAD5668-5G
3,4-Dimethyl-1H-pyrazole Phosphate ≥98.0% (by HPLC, titration analysis)
3,4-Dimethyl-1H-pyrazole Phosphate
Formula:
C₅H₁₁N₂O₄P MW: 194,13 g/mol |
Número MDL:
MFCD09842544 Núm. CAS: 202842-98-6 EINECS: 424-640-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 166.0 to 170.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...