3,4-Dihydro-5-hydroxy-1H-quinolin-2-one ≥98.0% (by total nitrogen basis)
Proveedor: TCI
Sinónimos:
5-Hydroxy-2-oxo-1,2,3,4-tetrahydroquinoline, 5-Hydroxy-3,4-dihydro-2(1H)-quinolinone
D5669-1G
D5669-5G
TCIAD5669-1GEA
58
EUR
TCIAD5669-1G
TCIAD5669-5G
3,4-Dihydro-5-hydroxy-1H-quinolin-2-one ≥98.0% (by total nitrogen basis)
3,4-Dihydro-5-hydroxy-1H-quinolin-2-one
Formula:
C₉H₉NO₂ MW: 163,18 g/mol |
Número MDL:
MFCD01862194 Núm. CAS: 30389-33-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 239.0 to 243.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...