4,7-Dichloro-1,10-phenanthroline ≥98.0% (by GC, titration analysis)
Proveedor: TCI
D5690-1G
D5690-200MG
TCIAD5690-1GEA
147
EUR
TCIAD5690-1G
TCIAD5690-200MG
4,7-Dichloro-1,10-phenanthroline ≥98.0% (by GC, titration analysis)
4,7-Dichloro-1,10-phenanthroline
Formula:
C₁₂H₆Cl₂N₂ MW: 249,09 g/mol |
Número MDL:
MFCD00233882 Núm. CAS: 5394-23-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystaline |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...