3,7-Dihidro-1,7-dimetil-1H-purina-2,6-diona ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
Paraxanthine, 1,7-Dimethyl-1H-purine-2,6(3H,7H)-dione
D5696-25MG
D5696-100MG
TCIAD5696-25MGEA
82
EUR
TCIAD5696-25MG
TCIAD5696-100MG
3,7-Dihidro-1,7-dimetil-1H-purina-2,6-diona ≥98.0% (by HPLC)
3,7-Dihidro-1,7-dimetil-1H-purina-2,6-diona
Formula:
C₇H₈N₄O₂ MW: 180,17 g/mol |
Número MDL:
MFCD00005727 Núm. CAS: 611-59-6 EINECS: 210-271-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 30.50 to 31.70 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...