Dimethyl 2,2'-Oxydiacetate ≥98.0% (por GC)
Proveedor: TCI
D5721-1G
D5721-5G
TCIAD5721-1GEA
77
EUR
TCIAD5721-1G
TCIAD5721-5G
Dimethyl 2,2'-Oxydiacetate ≥98.0% (por GC)
Dimethyl 2,2'-Oxydiacetate
Formula:
C₆H₁₀O₅ Pto. Ebullición: 126°C |
Número MDL:
MFCD01026217 Núm. CAS: 7040-23-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 36.0 to 40.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...