2,3-Diphenyl-1H-indole ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
D5741-1G
D5741-5G
TCIAD5741-1GEA
71
EUR
TCIAD5741-1G
TCIAD5741-5G
2,3-Diphenyl-1H-indole ≥98.0% (by HPLC, total nitrogen)
2,3-Diphenyl-1H-indole
Formula:
C₂₀H₁₅N MW: 269,35 g/mol |
Número MDL:
MFCD00022698 Núm. CAS: 3469-20-3 EINECS: 222-432-0 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...