Dibenzofuran-1-boronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: Dibenzo[b,d]furan-1-ylboronic acid

D5758-1G D5758-5G
TCIAD5758-1GEA 73 EUR
TCIAD5758-1G TCIAD5758-5G
Dibenzofuran-1-boronic acid (contains varying amounts of Anhydride)
Dibenzofuran-1-boronic acid
Formula: C₁₂H₉BO₃
Número MDL: MFCD01318978
Núm. CAS: 162607-19-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 95.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR