meso-Erythritol ≥99.0% (by HPLC, titration analysis)

Proveedor: TCI

E0021-25G E0021-500G E0021-100G
TCIAE0021-25GEA 18 EUR
TCIAE0021-25G TCIAE0021-500G TCIAE0021-100G
meso-Erythritol ≥99.0% (by HPLC, titration analysis)
meso-Erythritol
Formula: C₄H₁₀O₄
Número MDL: MFCD00004710
Núm. CAS: 149-32-6
EINECS: 205-737-3
Índice Merck: 14,03675

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 99.0 area%
Purity (Iodometric Titration) min. 99.0 %
Melting point 120.0 to 122.0 deg-C
Solubility in Water almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR