Esculina hidrato ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
Aesculin, 6,7-Dihydroxycoumarin 6-glucoside hydrate, Esculin
E0024-25G
E0024-5G
TCIAE0024-25GEA
219
EUR
TCIAE0024-25G
TCIAE0024-5G
Esculina hidrato ≥98.0% (by HPLC, titration analysis)
Esculina hidrato
Formula:
C₁₅H₁₆O₉·xH₂O MW: 340.29 g/mol (anhy) Pto. de fusión: 212 |
Número MDL:
MFCD00149492 Núm. CAS: 531-75-9 EINECS: 208-517-5 Índice Merck: 14,03698 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | -84.0 to -88.0 deg (C=2.4, Dioxane (50%)) (calcd.on anh.substance) |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...