Ethynylestradiol ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

E0037-1G E0037-5G
TCIAE0037-1GEA 51 EUR
TCIAE0037-1G TCIAE0037-5G
Ethynylestradiol ≥98.0% (by HPLC, titration analysis)
Ethynylestradiol
Formula: C₂₀H₂₄O₂
Número MDL: MFCD00003690
Núm. CAS: 57-63-6
EINECS: 200-342-2
Índice Merck: 14,03734

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 181.0 to 185.0 deg-C
Specific rotation [a]20/D -28.0 to -32.0 deg (C=0.4, Pyridene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR