Ethylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Aminoethane Hydrobromide
E0056-25G
E0056-500G
TCIAE0056-25GEA
16
EUR
TCIAE0056-25G
TCIAE0056-500G
Ethylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
Ethylamine hydrobromide
Formula:
C₂H₇N MW: 126,00 g/mol |
Número MDL:
MFCD00051999 Núm. CAS: 593-55-5 EINECS: 209-797-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 158.0 to 163.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...