Ethyl phthalyl ethyl glycolate ≥95.0% (por GC)

Proveedor: TCI

Sinónimos: Ethoxycarbonylmethyl Ethyl Phthalate, Phthalic Acid Ethoxycarbonylmethyl Ethyl Ester

E0164-25G E0164-500G
TCIAE0164-25GEA 16 EUR
TCIAE0164-25G TCIAE0164-500G
Ethyl phthalyl ethyl glycolate ≥95.0% (por GC)
Ethyl phthalyl ethyl glycolate
Formula: C₁₄H₁₆O₆
MW: 280,28 g/mol
Pto. de inflamación: 185°C
Número MDL: MFCD00059377
Núm. CAS: 84-72-0
EINECS: 201-555-3

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR