16α,17α-Epoxyprogesterone ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 16α-17α-Epoxy-δ4-pregnen-3,20-dione, 16α,17α-Epoxy-4-pregnen-3,20-dione, 16,17-Epoxyprogesterone

E0198-1G E0198-25G E0198-5G
TCIAE0198-1GEA 22 EUR
TCIAE0198-1G TCIAE0198-25G TCIAE0198-5G
16α,17α-Epoxyprogesterone ≥98.0% (by HPLC)
16α,17α-Epoxyprogesterone
Formula: C₂₁H₂₈O₃
MW: 328,452 g/mol
Número MDL: MFCD00055948
Núm. CAS: 1097-51-4
EINECS: 214-147-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +165.0 to +170.0 deg (C=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR