Sodium ethanesulphonate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

E0256-25G E0256-5G
TCIAE0256-25GEA 188 EUR
TCIAE0256-25G TCIAE0256-5G
Sodium ethanesulphonate ≥98.0% (by titrimetric analysis)
Sodium ethanesulphonate
Formula: C₂H₆O₃S
Número MDL: MFCD00066498
Núm. CAS: 5324-47-0
EINECS: 226-194-9

Order Now

Resultados test especificaciones

Appearance White powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Solubility in Water almost transparency
Drying loss max. 1.0 %
Haloid max. 0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR