1-Ethyl-2-phenylindole ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 1-Ethyl-2-phenyl-1H-indole

E0331-25G E0331-500G
TCIAE0331-25GEA 26 EUR
TCIAE0331-25G TCIAE0331-500G
1-Ethyl-2-phenylindole ≥98.0% (by HPLC)
1-Ethyl-2-phenylindole
Formula: C₁₆H₁₅N
MW: 221,30 g/mol
Pto. Ebullición: 209°C
Número MDL: MFCD00031481
Núm. CAS: 13228-39-2
EINECS: 236-199-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 83.0 to 87.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR