Ethyl-2-chloro-2-(hydroxyimino)acetate ≥99.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

E0371-25G E0371-5G
TCIAE0371-25GEA 126 EUR
TCIAE0371-25G TCIAE0371-5G
Ethyl-2-chloro-2-(hydroxyimino)acetate ≥99.0% (by titrimetric analysis)
Ethyl-2-chloro-2-(hydroxyimino)acetate
Formula: C₄H₆ClNO₃
Número MDL: MFCD00010209
Núm. CAS: 14337-43-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 99.0 %
Melting point 79.0 to 83.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR