2-Ethyl-2-methyl-1,3-propanediol ≥97.0% (por GC)

Proveedor: TCI

E0412-25G E0412-5G
TCIAE0412-25GEA 442 EUR
TCIAE0412-25G TCIAE0412-5G
2-Ethyl-2-methyl-1,3-propanediol ≥97.0% (por GC)
2-Ethyl-2-methyl-1,3-propanediol
Formula: C₆H₁₄O₂
Número MDL: MFCD00004693
Núm. CAS: 77-84-9
EINECS: 201-062-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %
Melting point 41.0 to 44.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR