(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: N-Ethyl-L-prolinamide

E0452-1G E0452-5G
TCIAE0452-1GEA 119 EUR
TCIAE0452-1G TCIAE0452-5G
(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide ≥98.0% (by GC, titration analysis)
(S)-(-)-1-Ethyl-2-pyrrolidinecarboxamide
Formula: C₇H₁₄N₂O
MW: 142,20 g/mol
Número MDL: MFCD00191361
Núm. CAS: 114812-34-9

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 112.0 deg-C
Specific rotation [a]20/D -114.0 to -120.0 deg (C=0.5, MeOH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR