3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
E0511-25G
E0511-5G
TCIAE0511-25GEA
69
EUR
TCIAE0511-25G
TCIAE0511-5G
3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide ≥98.0% (by HPLC, titration analysis)
3-Ethyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium bromide
Formula:
C₈H₁₄BrNOS |
Número MDL:
MFCD00040549 Núm. CAS: 54016-70-5 EINECS: 258-925-2 |
Resultados test especificaciones
Appearance | White to Light yellow to Light red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 83.0 to 88.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...