N-Ethylglycine ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
2-(Ethylamino)acetic acid, N-Et-Gly-OH
E0517-1G
E0517-25G
E0517-5G
TCIAE0517-1GEA
60
EUR
TCIAE0517-1G
TCIAE0517-25G
TCIAE0517-5G
N-Ethylglycine ≥98.0% (by titrimetric analysis)
N-Ethylglycine
Formula:
C₄H₉NO₂ MW: 103,12 g/mol |
Número MDL:
MFCD00037794 Núm. CAS: 627-01-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...