Ethyl-7-(diethylamino)coumarin-3-carboxylate ≥98.0% (by GC, titration analysis)
Proveedor: TCI
Sinónimos:
7-(Diethylamino)coumarin-3-carboxylic Acid Ethyl Ester, Ethyl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate
E0642-1G
E0642-5G
TCIAE0642-1GEA
59
EUR
TCIAE0642-1G
TCIAE0642-5G
Ethyl-7-(diethylamino)coumarin-3-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl-7-(diethylamino)coumarin-3-carboxylate
Formula:
C₁₆H₁₉NO₄ MW: 289,33 g/mol |
Número MDL:
MFCD00227484 Núm. CAS: 28705-46-6 |
Resultados test especificaciones
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 84.0 to 88.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...