(-)-Epigallocatechin gallate hydrate ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
EGCG hydrate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate hydrate, (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl gallate hydrate
E0694-100MG
E0694-500MG
TCIAE0694-100MGEA
39
EUR
TCIAE0694-100MG
TCIAE0694-500MG
(-)-Epigallocatechin gallate hydrate ≥98.0% (by HPLC)
(-)-Epigallocatechin gallate hydrate
Formula:
C₂₂H₁₈O₁₁ MW: 458,38 g/mol Pto. de fusión: 212°C |
Número MDL:
MFCD00075940 Núm. CAS: 989-51-5 EINECS: 479-560-7 Índice Merck: 14,03526 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -165.0 to -186.0 deg (C=1, EtOH) |
Water | max. 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...