3-Carbethoxy-2-piperidone ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: Ethyl 2-Piperidone-3-carboxylate, 2-Oxo-3-piperidinecarboxylic acid ethyl ester, Ethyl 2-Oxo-3-piperidinecarboxylate, 3-Ethoxycarbonyl-2-piperidone

E0742-25G E0742-5G
TCIAE0742-25GEA 121 EUR
TCIAE0742-25G TCIAE0742-5G
3-Carbethoxy-2-piperidone ≥98.0% (by HPLC)
3-Carbethoxy-2-piperidone
Formula: C8H13NO3
MW: 171.1900 g/mol
Número MDL: MFCD00006038
Núm. CAS: 3731-16-6
EINECS: 223-086-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 80.0 to 84.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR