1-Ethyl-2,3-dimethylimidazoliumbis(trifluoromethanesulphonyl)imide ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

Sinónimos: 1-EThyl-2,3-dimethylimidazolium bis(trifluoromethanesulphonyl)imide

E0753-25G E0753-5G
TCIAE0753-25GEA 298 EUR
TCIAE0753-25G TCIAE0753-5G
1-Ethyl-2,3-dimethylimidazoliumbis(trifluoromethanesulphonyl)imide ≥98.0% (by total nitrogen and titration analysis)
1-Ethyl-2,3-dimethylimidazoliumbis(trifluoromethanesulphonyl)imide
Formula: C₉H₁₃F₆N₃O₄S₂
MW: 405,33 g/mol
Número MDL: MFCD10566900
Núm. CAS: 174899-90-2

Order Now

Resultados test especificaciones

Purity (with Total Nitrogen) min. 98.0 %
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR