Ethyl-2-Amino-4-phenylthiazole-5-carboxylate ≥97.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
E0769-1G
E0769-5G
TCIAE0769-1GEA
45
EUR
TCIAE0769-1G
TCIAE0769-5G
Ethyl-2-Amino-4-phenylthiazole-5-carboxylate ≥97.0% (by HPLC, titration analysis)
Ethyl-2-Amino-4-phenylthiazole-5-carboxylate
Formula:
C₁₂H₁₂N₂O₂S |
Número MDL:
MFCD00022452 Núm. CAS: 64399-23-1 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 168.0 to 173.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...