Ethyl-3-amino-3-methylbutanoate hydrochloride ≥98.0% (by total nitrogen basis)

Proveedor: TCI

Sinónimos: Ethyl 3-amino-3-methylbutanoate hydrochloride

E0809-1G E0809-5G
TCIAE0809-1GEA 68 EUR
TCIAE0809-1G TCIAE0809-5G
Ethyl-3-amino-3-methylbutanoate hydrochloride ≥98.0% (by total nitrogen basis)
Ethyl-3-amino-3-methylbutanoate hydrochloride
Formula: C₇H₁₆ClNO₂
MW: 181,6604 g/mol
Número MDL: MFCD11844787
Núm. CAS: 85532-40-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Melting point 99.0 to 103.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR