Ethyl cyano(hydroxyimino)acetate ≥98.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: Ethyl 2-cyano-2-(hydroxyimino)acetate, Ethyl Cyanoglyoxalate Oxime, Cyano(hydroxyimino)acetic Acid Ethyl Ester, Oxyma

E0847-25G E0847-100G
TCIAE0847-25GEA 26 EUR
TCIAE0847-25G TCIAE0847-100G
Ethyl cyano(hydroxyimino)acetate ≥98.0% (por GC)
Ethyl cyano(hydroxyimino)acetate
Formula: C₅H₆N₂O₃
MW: 142,11 g/mol
Número MDL: MFCD00002112
Núm. CAS: 3849-21-6
EINECS: 223-351-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR