Entecavir monohydrate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-amino-9-((1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl)-1,9-dihydro-6H-purin-6-one hydrate, 9-[(1S,3R,4S)-4-Hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]guanine monohydrate
E0899-50MG
E0899-200MG
TCIAE0899-50MGEA
60
EUR
TCIAE0899-50MG
TCIAE0899-200MG
Entecavir monohydrate ≥98.0% (by HPLC, titration analysis)
Entecavir monohydrate
Formula:
C₁₂H₁₇N₅O₄ MW: 295,29 g/mol |
Número MDL:
MFCD09754448 Núm. CAS: 209216-23-9 Índice Merck: 14,03595 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % (calcd.on anh.substance) |
Specific rotation [a]20/D | +34.0 to +38.0 deg (C=0.25, H2O) (calcd.on anh.substance) |
Water | 4.0 to 9.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...