Ethyl-3-aminobenzofuran-2-carboxylate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Ethyl 3-Aminocoumarilate, 3-Aminobenzofuran-2-carboxylic acid Ethyl Ester, 3-Aminocoumarilic acid Ethyl Ester, Ethyl 3-aminobenzofuran-2-carboxylate
E0910-1G
E0910-5G
TCIAE0910-1GEA
25
EUR
TCIAE0910-1G
TCIAE0910-5G
Ethyl-3-aminobenzofuran-2-carboxylate ≥98.0% (por GC)
Ethyl-3-aminobenzofuran-2-carboxylate
Formula:
C₁₁H₁₁NO₃ MW: 205,21 g/mol |
Número MDL:
MFCD00641330 Núm. CAS: 39786-35-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 78.0 to 82.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...