Estra-5(10),9(11)-diene-3,17-dione-3-ethylene ketal ≥98.0% (by HPLC)

Proveedor: TCI
Peligro

E0918-1G E0918-5G
TCIAE0918-1GEA 31 EUR
TCIAE0918-1G TCIAE0918-5G
Estra-5(10),9(11)-diene-3,17-dione-3-ethylene ketal ≥98.0% (by HPLC)
Estra-5(10),9(11)-diene-3,17-dione-3-ethylene ketal
Formula: C₂₀H₂₆O₃
MW: 314,43 g/mol
Número MDL: MFCD08062585
Núm. CAS: 5571-36-8
EINECS: 427-230-8
UN: 3077
ADR: 9,III

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 154.0 to 158.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR