3-O-ethyl-L-ascorbic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: (R)-5-((S)-1,2-Dihydroxyethyl)-4-ethoxy-3-hydroxyfuran-2(5H)-one

E0926-5G E0926-25G
TCIAE0926-5GEA 53 EUR
TCIAE0926-5G TCIAE0926-25G
3-O-ethyl-L-ascorbic acid ≥98.0% (by HPLC, titration analysis)
3-O-ethyl-L-ascorbic acid
Formula: C₈H₁₂O₆
MW: 204,18 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD09261382
Núm. CAS: 86404-04-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 112.0 to 116.0 deg-C
Specific rotation [a]20/D +45.0 to +49.0 deg (C=1、MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR