Ethyl-1-aminocyclopropanecarboxylate hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Proveedor: TCI
Atención

Sinónimos: H-Acpc-OEt.HCl, H-Acpc-OEt·HCl

E0937-1G E0937-5G
TCIAE0937-1GEA 24 EUR
TCIAE0937-1G TCIAE0937-5G
Ethyl-1-aminocyclopropanecarboxylate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Ethyl-1-aminocyclopropanecarboxylate hydrochloride
Formula: C₆H₁₁NO₂·HCl
MW: 165,6 g/mol
Número MDL: MFCD00190747
Núm. CAS: 42303-42-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point gt. 110.0 and le 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR