Ethyl-2-aminoisonicotinate ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Amino-isonicotinic acid ethyl ester, Ethyl 2-aminopyridine-4-carboxylate, 2-Aminopyridine-4-carboxylic acid Ethyl Ester, Ethyl 2-aminoisonicotinate, Ethyl 2-Aminopyridine-4-carboxylate, 2-Aminoisonicotinic acid Ethyl Ester, Ethyl 2-Aminoisonicotinate
E0944-1G
E0944-5G
TCIAE0944-1GEA
25
EUR
TCIAE0944-1G
TCIAE0944-5G
Ethyl-2-aminoisonicotinate ≥98.0% (by GC, titration analysis)
Ethyl 2-aminoisonicotinate
Formula:
C₈H₁₀N₂O₂ MW: 166,18 g/mol |
Número MDL:
MFCD03791260 Núm. CAS: 13362-30-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 123.0 to 127.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...