1-Boc-DL-nipecotic acid ethyl ester ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Ethyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate, Ethyl 1-Boc-DL-nipecotate, Ethyl 1-Boc-3-piperidinecarboxylate
E0954-5G
E0954-25G
TCIAE0954-5GEA
42
EUR
TCIAE0954-5G
TCIAE0954-25G
1-Boc-DL-nipecotic acid ethyl ester ≥98.0% (por GC)
1-Boc-DL-nipecotic acid ethyl ester
Formula:
C₁₃H₂₃NO₄ MW: 257,33 g/mol |
Número MDL:
MFCD04116274 Núm. CAS: 130250-54-3 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 34.0 to 38.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...