Ethyl-2-aminobenzo[d]thiazole-6-carboxylate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

E0985-1G E0985-5G
TCIAE0985-1GEA 23 EUR
TCIAE0985-1G TCIAE0985-5G
Ethyl-2-aminobenzo[d]thiazole-6-carboxylate ≥98.0% (by HPLC, titration analysis)
Ethyl-2-aminobenzo[d]thiazole-6-carboxylate
Formula: C₁₀H₁₀N₂O₂S
MW: 222,26 g/mol
Número MDL: MFCD00102724
Núm. CAS: 50850-93-6

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR